missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)benzyl bromide, 97%

Catalog No. p-7029342
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2679503 1 g
AAH2679506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2679503 Supplier Thermo Scientific Chemicals Supplier No. H2679503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86256-24-8 | C8H5BrF4O | 273.025 g/mol

2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 86256-24-8
Molecular Formula C8H5BrF4O
Molecular Weight (g/mol) 273.025
MDL Number MFCD04115890
InChI Key KOISGBFWQAPLFE-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethoxy benzyl bromide, 2-bromomethyl-1-fluoro-4-trifluoromethoxy benzene, 1-bromomethyl-2-fluoro-5-trifluoromethoxy benzene
PubChem CID 2783326
IUPAC Name 2-(bromomethyl)-1-fluoro-4-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)(F)F)CBr)F

Specifications

Refractive Index 1.466
Quantity 1 g
UN Number UN3265
Solubility Information Difficult to mix.
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only