missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7129646
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03735G 5 g
F037325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03735G Supplier TCI America Supplier No. F03735G
Only null left

Chemical Identifiers

CAS 400-74-8
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00024576
InChI Key DNTHMWUMRGOJRY-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrobenzotrifluoride, 1-fluoro-4-nitro-2-trifluoromethyl benzene, benzene, 1-fluoro-4-nitro-2-trifluoromethyl, alpha,alpha,alpha-2-tetrafluorotoluene, 2-fluoro-5-nitro trifluoromethyl benzene, 1-fluoro-4-nitro-2-trifluoromethyl-benzene, 1-fluoro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-alpha,alpha,alpha,2-tetrafluorotoluene, pubchem1653, 2-fluor-5-nitrobenzotrifluorid
PubChem CID 94954
IUPAC Name 1-fluoro-4-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)F

Specifications

Boiling Point 110°C
Quantity 5 g
Formula Weight 209.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-nitrobenzotrifluoride