accessibility menu, dialog, popup

UserName

CAS RN 400-74-8

IUPAC Name: 1-fluoro-4-nitro-2-(trifluoromethyl)benzene
Synonyms: 1-fluoro-4-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 209.1
Molecular Formula: C7H3F4NO2
InChi Key: DNTHMWUMRGOJRY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)