missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzotrifluoride, 98%

Catalog No. AAA1813006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1813006 5 g
AAA1813014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1813006 Supplier Thermo Scientific Chemicals Supplier No. A1813006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 400-74-8 | C7H3F4NO2 | 209.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 400-74-8
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00024576
InChI Key DNTHMWUMRGOJRY-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrobenzotrifluoride, 1-fluoro-4-nitro-2-trifluoromethyl benzene, benzene, 1-fluoro-4-nitro-2-trifluoromethyl, alpha,alpha,alpha-2-tetrafluorotoluene, 2-fluoro-5-nitro trifluoromethyl benzene, 1-fluoro-4-nitro-2-trifluoromethyl-benzene, 1-fluoro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-alpha,alpha,alpha,2-tetrafluorotoluene, pubchem1653, 2-fluor-5-nitrobenzotrifluorid
PubChem CID 94954
IUPAC Name 1-fluoro-4-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)F

Specifications

Density 1.522
Boiling Point 105°C to 110°C (25 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.465
Quantity 5 g
UN Number UN2810
Beilstein 1881423
Formula Weight 209.1
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzotrifluoride
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-925-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only