missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylbenzonitrile 98.0+%, TCI America™

Catalog No. F05735G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F05735G Supplier TCI America Supplier No. F05735G

Chemical Identifiers

CAS 64113-84-4
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00134542
InChI Key CMAOLVNGLTWICC-UHFFFAOYSA-N
Synonym 6-fluoro-m-tolunitrile, benzonitrile, 2-fluoro-5-methyl, 2-fluoro-5-methyl-benzonitrile, 3-cyano-4-fluorotoluene, 2-fluoro-5-methylbenzenecarbonitrile, pubchem1552, acmc-209nk8, ksc352m4h, 2-fluoro-5 methyl benzonitrile, 2-fluoro-5-methyl benzonitrile
PubChem CID 2774584
IUPAC Name 2-fluoro-5-methylbenzonitrile
SMILES CC1=CC(=C(C=C1)F)C#N

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 5 g
Formula Weight 135.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-methylbenzonitrile