accessibility menu, dialog, popup

UserName

CH3FN

CAS RN 64113-84-4

IUPAC Name: 2-fluoro-5-methylbenzonitrile
Synonyms: 2-fluoro-5-methylbenzonitrile
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: CMAOLVNGLTWICC-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(=C1)C#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)