missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylbenzonitrile, 99%

Catalog No. AAH2616603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2616603 1 g
AAH2616606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2616603 Supplier Thermo Scientific Chemicals Supplier No. H2616603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 64113-84-4 | C8H6FN | 135.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64113-84-4
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00134542
InChI Key CMAOLVNGLTWICC-UHFFFAOYSA-N
Synonym 6-fluoro-m-tolunitrile, benzonitrile, 2-fluoro-5-methyl, 2-fluoro-5-methyl-benzonitrile, 3-cyano-4-fluorotoluene, 2-fluoro-5-methylbenzenecarbonitrile, pubchem1552, acmc-209nk8, ksc352m4h, 2-fluoro-5 methyl benzonitrile, 2-fluoro-5-methyl benzonitrile
PubChem CID 2774584
IUPAC Name 2-fluoro-5-methylbenzonitrile
SMILES CC1=CC(=C(C=C1)F)C#N

Specifications

Melting Point 49°C to 52°C
Flash Point 94°C (201°F)
Quantity 1 g
UN Number UN3439
Formula Weight 135.14
Percent Purity 99%
Chemical Name or Material 2-Fluoro-5-methylbenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only