missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzonitrile 98.0+%, TCI America™

Catalog No. F06425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06425G 5 g
F06421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06425G Supplier TCI America Supplier No. F06425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127667-01-0
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00142921
InChI Key VBZLRHYLNXWZIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-benzonitrile, benzonitrile, 2-fluoro-5-methoxy, 3-cyano-4-fluoroanisole, fluoromethoxybenzonitrile2, 2-fluoro-5-methoxybenzenecarbonitrile, pubchem3500, acmc-209bct, ksc494m9f, 2-fluoro-5-methoxy benzonitrile, buttpark 80\01-21
PubChem CID 2774546
IUPAC Name 2-fluoro-5-methoxybenzonitrile
SMILES COC1=CC(C#N)=C(F)C=C1

Specifications

Melting Point 82°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 151.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-methoxybenzonitrile