accessibility menu, dialog, popup

UserName

H3COFN

CAS RN 127667-01-0

IUPAC Name: 2-fluoro-5-methoxybenzonitrile
Synonyms: 2-fluoro-5-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: VBZLRHYLNXWZIU-UHFFFAOYSA-N
SMILES: COC1=CC(C#N)=C(F)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)