missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

Catalog No. p-7031595
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3272406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3272406 Supplier Thermo Scientific Chemicals Supplier No. H3272406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127667-01-0
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00142921
InChI Key VBZLRHYLNXWZIU-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-benzonitrile, benzonitrile, 2-fluoro-5-methoxy, 3-cyano-4-fluoroanisole, fluoromethoxybenzonitrile2, 2-fluoro-5-methoxybenzenecarbonitrile, pubchem3500, acmc-209bct, ksc494m9f, 2-fluoro-5-methoxy benzonitrile, buttpark 80\01-21
PubChem CID 2774546
IUPAC Name 2-fluoro-5-methoxybenzonitrile
SMILES COC1=CC(C#N)=C(F)C=C1

Specifications

Melting Point 81°C to 85°C
Quantity 5 g
UN Number UN3439
Formula Weight 151.14
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-methoxybenzonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only