missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
Supplier: TCI America F10945G
Specifications
| 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 31°C | |
| C11H15BFNO2 | |
| 5 g | |
| ZKSRQMCKFLGPQU-UHFFFAOYSA-N | |
| 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
| 12177222 | |
| ≥98.0% (GC,T) |
| 444120-95-0 | |
| White-Yellow | |
| MFCD03412792 | |
| 2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Fluoropyridine-3-boronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)F | |
| 223.054 | |
| 223.05 | |
| Crystalline Lumps |
Chemical Identifiers
| 444120-95-0 | |
| 223.054 | |
| ZKSRQMCKFLGPQU-UHFFFAOYSA-N | |
| 12177222 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)F |
| C11H15BFNO2 | |
| MFCD03412792 | |
| 2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Fluoropyridine-3-boronic Acid Pinacol Ester | |
| 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator