missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7130064
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10941G 1 g
F10945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10941G Supplier TCI America Supplier No. F10941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 444120-95-0 | C11H15BFNO2 | 223.054 g/mol

Chemical Identifiers

CAS 444120-95-0
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.054
MDL Number MFCD03412792
InChI Key ZKSRQMCKFLGPQU-UHFFFAOYSA-N
Synonym 2-(6-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Fluoropyridine-3-boronic Acid Pinacol Ester
PubChem CID 12177222
IUPAC Name 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)F

Specifications

Melting Point 31°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
TSCA No
Recommended Storage Refrigerator