accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNF

CAS RN 444120-95-0

IUPAC Name: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 223.05
Molecular Formula: C11H15BFNO2
InChi Key: ZKSRQMCKFLGPQU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results