missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%

Catalog No. AAB2048506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2048506 5 g
AAB2048514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2048506 Supplier Thermo Scientific Chemicals Supplier No. B2048506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 146070-34-0 | C8H3F4N | 189.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146070-34-0
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00061279
InChI Key JLTYVTXTSOYXMX-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl benzonitrile, 3-fluoro-4-cyano-benzotrifluoride, 4-cyano-3-fluorobenzotrifluoride, 2-fluoro-4-trifluoromethyl-benzonitrile, benzonitrile, 2-fluoro-4-trifluoromethyl, alpha,alpha,alpha,2-tetrafluoro-p-tolunitrile, 2-fluoro-4-trifluoromethyl benzenecarbonitrile, .alpha.,.alpha.,.alpha.,2-tetrafluoro-p-tolunitrile, pubchem4798, intermediates-zcf02116
PubChem CID 518973
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C#N

Specifications

Density 1.358
Boiling Point 178°C to 180°C
Flash Point 75°C (167°F)
Refractive Index 1.446
Quantity 5 g
UN Number UN3276
Beilstein 8313354
Formula Weight 189.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzonitrile
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only