accessibility menu, dialog, popup

UserName

CAS RN 146070-34-0

IUPAC Name: 2-fluoro-4-(trifluoromethyl)benzonitrile
Synonyms: 2-fluoro-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: JLTYVTXTSOYXMX-UHFFFAOYSA-N
SMILES: FC1=CC(=CC=C1C#N)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results