Learn More
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
CAS: 89763-93-9 | C8H4F4O | 192.113 g/mol
Supplier: Thermo Scientific Chemicals A1850014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Fluoro-4-(trifluoromethyl)benzaldehyde | |
| 1.41 | |
| 110° (230°F) | |
| 1.45 | |
| 25 g | |
| Air Sensitive | |
| KFEHNXLFIGPWNB-UHFFFAOYSA-N | |
| 2-fluoro-4-(trifluoromethyl)benzaldehyde | |
| 522929 | |
| 97% |
| 89763-93-9 | |
| 118°C to 119°C | |
| C8H4F4O | |
| MFCD00061310 | |
| 7703632 | |
| 2-fluoro-4-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-4-trifluoromethyl, pubchem4235, acmc-209r2s, ksc496e9p, attercop-chm at111109, a,a,a,2-tetrafluoro-p-tolualdehyde, 2-fluoro-4-trifluoromethyl benzaldehy, 2-fluoro-4-trifluoromethyl-benzaldehyde, 2-fluoro4-trifluoromethyl benzaldehyde | |
| C1=CC(=C(C=C1C(F)(F)F)F)C=O | |
| 192.113 | |
| 192.11 |
Chemical Identifiers
| 89763-93-9 | |
| 192.113 | |
| KFEHNXLFIGPWNB-UHFFFAOYSA-N | |
| 522929 | |
| C1=CC(=C(C=C1C(F)(F)F)F)C=O |
| C8H4F4O | |
| MFCD00061310 | |
| 2-fluoro-4-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-4-trifluoromethyl, pubchem4235, acmc-209r2s, ksc496e9p, attercop-chm at111109, a,a,a,2-tetrafluoro-p-tolualdehyde, 2-fluoro-4-trifluoromethyl benzaldehy, 2-fluoro-4-trifluoromethyl-benzaldehyde, 2-fluoro4-trifluoromethyl benzaldehyde | |
| 2-fluoro-4-(trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only