CAS RN 89763-93-9
IUPAC Name:
2-fluoro-4-(trifluoromethyl)benzaldehyde
Synonyms:
2-fluoro-4-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
192.11
Molecular Formula:
C8H4F4O
InChi Key:
KFEHNXLFIGPWNB-UHFFFAOYSA-N
SMILES:
FC1=CC(=CC=C1C=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%
CAS: 89763-93-9 Molecular Formula: C8H4F4O Molecular Weight (g/mol): 192.113 MDL Number: MFCD00061310 InChI Key: KFEHNXLFIGPWNB-UHFFFAOYSA-N Synonym: 2-fluoro-4-trifluoromethyl benzaldehyde,benzaldehyde, 2-fluoro-4-trifluoromethyl,pubchem4235,acmc-209r2s,ksc496e9p,attercop-chm at111109,a,a,a,2-tetrafluoro-p-tolualdehyde,2-fluoro-4-trifluoromethyl benzaldehy,2-fluoro-4-trifluoromethyl-benzaldehyde,2-fluoro4-trifluoromethyl benzaldehyde PubChem CID: 522929 IUPAC Name: 2-fluoro-4-(trifluoromethyl)benzaldehyde SMILES: C1=CC(=C(C=C1C(F)(F)F)F)C=O