missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%

Catalog No. AAA1850003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1850003 1 g
AAA1850006 5 g
AAA1850014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1850003 Supplier Thermo Scientific Chemicals Supplier No. A1850003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89763-93-9 | C8H4F4O | 192.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89763-93-9
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061310
InChI Key KFEHNXLFIGPWNB-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-4-trifluoromethyl, pubchem4235, acmc-209r2s, ksc496e9p, attercop-chm at111109, a,a,a,2-tetrafluoro-p-tolualdehyde, 2-fluoro-4-trifluoromethyl benzaldehy, 2-fluoro-4-trifluoromethyl-benzaldehyde, 2-fluoro4-trifluoromethyl benzaldehyde
PubChem CID 522929
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C=O

Specifications

Density 1.41
Boiling Point 118°C to 119°C
Flash Point 110° (230°F)
Refractive Index 1.45
Quantity 1 g
Beilstein 7703632
Sensitivity Air Sensitive
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only