missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4-pentyl-4″-propyl-1,1':4',1″-terphenyl 98.0+%, TCI America™

Catalog No. p-7130046
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10461G 1 g
F10465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10461G Supplier TCI America Supplier No. F10461G
Only null left

Chemical Identifiers

CAS 95759-51-6
Molecular Formula C26H29F
Molecular Weight (g/mol) 360.516
MDL Number MFCD11045061
InChI Key SXGOKAUBXXCAAC-UHFFFAOYSA-N
PubChem CID 17977863
IUPAC Name 2-fluoro-1-(4-pentylphenyl)-4-(4-propylphenyl)benzene
SMILES CCCCCC1=CC=C(C=C1)C2=C(C=C(C=C2)C3=CC=C(C=C3)CCC)F

Specifications

Melting Point 53°C
Color White
Quantity 1 g
Synonym 2′C-Fluoro-4-pentyl-4′C′C-propyl-p-terphenyl
Formula Weight 360.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Fluoro-4-pentyl-4′′-propyl-1,1′:4′,1′′-terphenyl