accessibility menu, dialog, popup

UserName

CAS RN 95759-51-6

IUPAC Name: 3-fluoro-4-(4-pentylphenyl)-4'-propyl-1,1'-biphenyl
Synonyms: 3-fluoro-4-(4-pentylphenyl)-4'-propyl-1,1'-biphenyl
Molecular Weight (g/mol): 360.52
Molecular Formula: C26H29F
InChi Key: SXGOKAUBXXCAAC-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(C=C1)C1=C(F)C=C(C=C1)C1=CC=C(CCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results