Learn More
2-Fluoro-4-nitrotoluene, 98%
CAS: 1427-07-2 | C7H6FNO2 | 155.128 g/mol
Supplier: Thermo Scientific Chemicals A1446422
Description
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 2-Fluoro-4-nitrotoluene | |
| 32°C to 37°C | |
| 73°C (163°F) | |
| MFCD00007199 | |
| UN2811 | |
| 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol | |
| WIQISTBTOQNVCE-UHFFFAOYSA-N | |
| 2-fluoro-1-methyl-4-nitrobenzene | |
| 74025 | |
| 98% |
| 1427-07-2 | |
| 56°C to 58°C (1 mmHg) | |
| C7H6FNO2 | |
| 100 g | |
| 2250156 | |
| Insoluble in water. Solubility in methanol gives very faint turbidity. | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])F | |
| 155.128 | |
| 155.13 |
Chemical Identifiers
| 1427-07-2 | |
| 155.128 | |
| WIQISTBTOQNVCE-UHFFFAOYSA-N | |
| 74025 | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])F |
| C7H6FNO2 | |
| MFCD00007199 | |
| 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol | |
| 2-fluoro-1-methyl-4-nitrobenzene |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 215-845-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only