missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrotoluene, 98%

Catalog No. p-7021929
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1446406 5 g
AAA1446414 25 g
AAA1446422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1446406 Supplier Thermo Scientific Chemicals Supplier No. A1446406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1427-07-2 | C7H6FNO2 | 155.128 g/mol

It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1427-07-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007199
InChI Key WIQISTBTOQNVCE-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
PubChem CID 74025
IUPAC Name 2-fluoro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

Melting Point 32°C to 37°C
Boiling Point 56°C to 58°C (1 mmHg)
Flash Point 73°C (163°F)
Quantity 5 g
UN Number UN2811
Beilstein 2250156
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity.
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 215-845-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only