missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylaniline 97.0+%, TCI America™

Catalog No. p-7129733
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05295G 5 g
F052925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05295G Supplier TCI America Supplier No. F05295G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 452-80-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00040975
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
PubChem CID 67984
IUPAC Name 2-fluoro-4-methylaniline
SMILES CC1=CC=C(N)C(F)=C1

Specifications

Melting Point 3°C
Boiling Point 71°C
Quantity 5 g
UN Number 2810
Formula Weight 125.15
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-methylaniline
RTECSNumber XU6650000
TSCA No