accessibility menu, dialog, popup

UserName

CAS RN 452-80-2

IUPAC Name: 2-fluoro-4-methylaniline
Synonyms: 2-fluoro-4-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: ZQEXBVHABAJPHJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(F)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)