missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylaniline, 99%

Catalog No. AAA1659106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1659106 5 g
AAA1659114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1659106 Supplier Thermo Scientific Chemicals Supplier No. A1659106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 452-80-2 | C7H8FN | 125.15 g/mol

2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

Solubility
Not miscible in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 452-80-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00040975
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
PubChem CID 67984
IUPAC Name 2-fluoro-4-methylaniline
SMILES CC1=CC=C(N)C(F)=C1

Specifications

Density 1.108
Boiling Point 70°C to 71°C (7 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.533
Quantity 5 g
UN Number UN2810
Beilstein 2637578
Solubility Information Not miscible in water.
Formula Weight 125.15
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-methylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber XU6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only