missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%

Catalog No. p-7040098
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59738MD 250 mg
AAH5973803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59738MD Supplier Thermo Scientific Chemicals Supplier No. H59738MD
Only null left

CAS: 1259022-70-2 | C15H20BFO4 | 294.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1259022-70-2
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.13
MDL Number MFCD18427635
InChI Key ZFLOTGNTGSGUGD-UHFFFAOYSA-N
Synonym methyl 2-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-3-fluoro-4-4,4,5 5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate
PubChem CID 59627674
IUPAC Name methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
SMILES COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Formula Weight 294.13
Percent Purity 96%
Chemical Name or Material 2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only