accessibility menu, dialog, popup

UserName

CAS RN 1259022-70-2

BOOH3CCH3H3CH3COOCH3F

CAS RN 1259022-70-2

IUPAC Name: methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
Synonyms: methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
Molecular Weight (g/mol): 294.13
Molecular Formula: C15H20BFO4
InChi Key: ZFLOTGNTGSGUGD-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results