missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodobenzonitrile, 98%

Catalog No. AAH6197306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197306 5 g
AAH6197303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197306 Supplier Thermo Scientific Chemicals Supplier No. H6197306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 137553-42-5 | C7H3FIN | 247.011 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 137553-42-5
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.011
MDL Number MFCD03094172
InChI Key WHQVXHBSTRFRCE-UHFFFAOYSA-N
Synonym 3-fluoro-4-cyano-1-iodobenzene, benzonitrile, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenecarbonitrile, pubchem4778, acmc-209xcp, intermediates-zcf02061, 4-iodo-2-fluorobenzonitrile, 2-fluoro-4-iodo-benzonitrile, ksc493i5t, benzonitrile,2-fluoro-4-iodo
PubChem CID 2774518
IUPAC Name 2-fluoro-4-iodobenzonitrile
SMILES C1=CC(=C(C=C1I)F)C#N

Specifications

Color Red to Brown
Quantity 5 g
UN Number UN3439
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 247.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only