accessibility menu, dialog, popup

UserName

IFN

CAS RN 137553-42-5

IUPAC Name: 2-fluoro-4-iodobenzonitrile
Synonyms: 2-fluoro-4-iodobenzonitrile
Molecular Weight (g/mol): 247.01
Molecular Formula: C7H3FIN
InChi Key: WHQVXHBSTRFRCE-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(I)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results