missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-formylbenzonitrile 97.0+%, TCI America™

Catalog No. F11121G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F11121G Supplier TCI America Supplier No. F11121G

Chemical Identifiers

CAS 101048-76-4
Molecular Formula C8H4FNO
Molecular Weight (g/mol) 149.12
MDL Number MFCD06738989
InChI Key MYUPCEIJNBAAFL-UHFFFAOYSA-N
Synonym 4-Cyano-3-fluorobenzaldehyde
PubChem CID 11217339
IUPAC Name 2-fluoro-4-formylbenzonitrile
SMILES FC1=C(C=CC(C=O)=C1)C#N

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 149.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-formylbenzonitrile