accessibility menu, dialog, popup

UserName

OFN

CAS RN 101048-76-4

IUPAC Name: 2-fluoro-4-formylbenzonitrile
Synonyms: 2-fluoro-4-formylbenzonitrile
Molecular Weight (g/mol): 149.12
Molecular Formula: C8H4FNO
InChi Key: MYUPCEIJNBAAFL-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(C=O)=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results