missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. F11251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11251G 1 g
F11255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11251G Supplier TCI America Supplier No. F11251G

Chemical Identifiers

CAS 146070-35-1
Molecular Formula C8H3F4N
Molecular Weight (g/mol) 189.113
MDL Number MFCD00061152
InChI Key NMWOWGXPPBIZNH-UHFFFAOYSA-N
Synonym 2-fluoro-3-trifluoromethyl benzonitrile, 3-cyano-2-fluorobenzotrifluoride, .alpha.,.alpha.,.alpha.,2-tetrafluoro-m-tolunitrile, 2-fluoro-3-trifluoromethyl benzenecarbonitrile, acmc-20anvp, buttpark 45\01-75, 3-trifluoromethyl-2-fluorobenzonitrile, 2-fluoro-3-trifluoromethyl-benzonitrile
PubChem CID 518974
IUPAC Name 2-fluoro-3-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C(=C1)C(F)(F)F)F)C#N

Specifications

Color Yellow
Quantity 1 g
UN Number 3276
Formula Weight 189.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzonitrile