accessibility menu, dialog, popup

UserName

FFFFN

CAS RN 146070-35-1

IUPAC Name: 2-fluoro-3-(trifluoromethyl)benzonitrile
Synonyms: 2-fluoro-3-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: NMWOWGXPPBIZNH-UHFFFAOYSA-N
SMILES: FC1=C(C=CC=C1C(F)(F)F)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results