missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals

$463.10 - $1,281.34

Chemical Identifiers

CAS 80049-87-2
Molecular Formula C10H11FN4O4
Molecular Weight (g/mol) 270.22
MDL Number MFCD09750860
InChI Key NRVOTDBYJXFINS-GPGUAWMRNA-N
PubChem CID 196148
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65208MD
View Documents Promotion Details
N/A 250 mg
Each for $463.10
Only null left
 
AAJ6520803
View Documents Promotion Details
N/A 1 g
Each for $1,281.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80049-87-2
270.22
NRVOTDBYJXFINS-GPGUAWMRNA-N
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
C10H11FN4O4
MFCD09750860
196148

Specifications

80049-87-2
C10H11FN4O4
2'-fluoro-2'-deoxyinosine, 2'-deoxy-2'-fluoroinosine, inosine,2'-deoxy-2'-fluoro, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2'-dfin, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, 2'-deoxy-2'-fluoro-inosine, inosine, 2'-deoxy-2'-fluoro, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl hypoxanthine, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purin-6 1h-one
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
270.22
270.22
Powder
White
MFCD09750860
NRVOTDBYJXFINS-GPGUAWMRNA-N
9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
196148
99%
2'-Fluoro-2'-deoxyinosine
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only