missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals

Catalog No. AAJ65208MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65208MD 250 mg
AAJ6520803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65208MD Supplier Thermo Scientific Chemicals Supplier No. J65208MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80049-87-2 | Molecular Formula: C10H11FN4O4 | Molecular Weight: 270.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80049-87-2
Molecular Formula C10H11FN4O4
Molecular Weight (g/mol) 270.22
MDL Number MFCD09750860
InChI Key NRVOTDBYJXFINS-GPGUAWMRNA-N
PubChem CID 196148
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Color White
Quantity 250 mg
Synonym 2'-fluoro-2'-deoxyinosine, 2'-deoxy-2'-fluoroinosine, inosine,2'-deoxy-2'-fluoro, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2'-dfin, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, 2'-deoxy-2'-fluoro-inosine, inosine, 2'-deoxy-2'-fluoro, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl hypoxanthine, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purin-6 1h-one
IUPAC Name 9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Formula Weight 270.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyinosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only