missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyinosine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J6520803

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6520803


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80049-87-2
270.22
NRVOTDBYJXFINS-GPGUAWMRNA-N
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
C10H11FN4O4
MFCD09750860
196148

Specifications

2'-Fluoro-2'-deoxyinosine
White
1 g
2'-fluoro-2'-deoxyinosine, 2'-deoxy-2'-fluoroinosine, inosine,2'-deoxy-2'-fluoro, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2'-dfin, 9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, 2'-deoxy-2'-fluoro-inosine, inosine, 2'-deoxy-2'-fluoro, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl hypoxanthine, 9-2-deoxy-2-fluoro-beta-d-ribofuranosyl-9h-purin-6 1h-one
OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=NC2=C1N=CNC2=O
270.22
270.22
Powder
80049-87-2
C10H11FN4O4
MFCD09750860
NRVOTDBYJXFINS-GPGUAWMRNA-N
9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
196148
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions