Learn More
Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyguanosine, 99%
CAS: 78842-13-4 | C10H12FN5O4 | 285.24 g/mol
Supplier: Thermo Scientific Chemicals J6405503
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 78842-13-4 | |
| 285.24 | |
| UXUZARPLRQRNNX-SJLGBTOWNA-N | |
| 196536 | |
| NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1 |
| C10H12FN5O4 | |
| MFCD00923832 | |
| 2'-deoxy-2'-fluoroguanosine, 2'-fluoro-2'-deoxyguanosine, guanosine, 2'-deoxy-2'-fluoro, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl-1,9-dihydro-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2'-dfgs, deoxyfluoroguanosine, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one | |
| 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one |
Specifications
| 2'-Fluoro-2'-deoxyguanosine | |
| White | |
| 1 g | |
| 2'-deoxy-2'-fluoroguanosine, 2'-fluoro-2'-deoxyguanosine, guanosine, 2'-deoxy-2'-fluoro, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl-1,9-dihydro-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2'-dfgs, deoxyfluoroguanosine, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one | |
| NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1 | |
| 285.24 | |
| 285.23 | |
| Powder |
| 78842-13-4 | |
| C10H12FN5O4 | |
| MFCD00923832 | |
| UXUZARPLRQRNNX-SJLGBTOWNA-N | |
| 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one | |
| 196536 | |
| 99% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold