missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyguanosine, 99%

Catalog No. p-7042866
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64055MD 250 mg
AAJ6405503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64055MD Supplier Thermo Scientific Chemicals Supplier No. J64055MD

CAS: 78842-13-4 | C10H12FN5O4 | 285.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78842-13-4
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.24
MDL Number MFCD00923832
InChI Key UXUZARPLRQRNNX-SJLGBTOWNA-N
Synonym 2'-deoxy-2'-fluoroguanosine, 2'-fluoro-2'-deoxyguanosine, guanosine, 2'-deoxy-2'-fluoro, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl-1,9-dihydro-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2'-dfgs, deoxyfluoroguanosine, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one
PubChem CID 196536
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1

Specifications

Color White
Quantity 250 mg
Formula Weight 285.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyguanosine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only