missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-1,4-dimethoxybenzene, 97%, Thermo Scientific Chemicals

$208.30 - $208.30

Chemical Identifiers

CAS 82830-49-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042487
InChI Key WNCYZVMZKSOPMU-UHFFFAOYSA-N
Synonym 1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
PubChem CID 145002
IUPAC Name 2-fluoro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1350506
View Documents
Thermo Scientific Chemicals
L1350506
5 g
Each for $208.30
Only null left
 
Description

Description

2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

82830-49-7
156.156
WNCYZVMZKSOPMU-UHFFFAOYSA-N
145002
COC1=CC(=C(C=C1)OC)F
C8H9FO2
MFCD00042487
1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
2-fluoro-1,4-dimethoxybenzene

Specifications

82830-49-7
119°C to 121°C (40 mmHg)
1.5045
5 g
1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
WNCYZVMZKSOPMU-UHFFFAOYSA-N
2-fluoro-1,4-dimethoxybenzene
145002
97%
23°C to 26°C
C8H9FO2
MFCD00042487
6387735
Insoluble in water.
COC1=CC(=C(C=C1)OC)F
156.156
156.16
2-Fluoro-1,4-dimethoxybenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only