accessibility menu, dialog, popup

UserName

H3COOCH3F

CAS RN 82830-49-7

IUPAC Name: 2-fluoro-1,4-dimethoxybenzene
Synonyms: 2-fluoro-1,4-dimethoxybenzene
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: WNCYZVMZKSOPMU-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC)C(F)=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)