Learn More
2-Ethylphenylhydrazine hydrochloride, 97%
CAS: 19398-06-2 | C8H12N2 | 136.20 g/mol
$395.42 - $1292.21
Chemical Identifiers
| CAS | 19398-06-2 |
|---|---|
| Molecular Formula | C8H12N2 |
| Molecular Weight (g/mol) | 136.20 |
| MDL Number | MFCD00071599 |
| InChI Key | JHPOWXCLWLEKBY-UHFFFAOYSA-N |
| Synonym | 2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride |
| PubChem CID | 2733209 |
| SMILES | CCC1=CC=CC=C1NN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1685318
|
Thermo Scientific Chemicals
A1685318 |
50 g |
Each for $395.42
|
|
|||||
|
AAA1685330
|
Thermo Scientific Chemicals
A1685330 |
250 g |
Each for $1,292.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19398-06-2 | |
| 136.20 | |
| JHPOWXCLWLEKBY-UHFFFAOYSA-N | |
| 2733209 |
| C8H12N2 | |
| MFCD00071599 | |
| 2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride | |
| CCC1=CC=CC=C1NN |
Specifications
| 19398-06-2 | |
| 1.21 | |
| MFCD00071599 | |
| UN3077 | |
| 2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride | |
| CCC1=CC=CC=C1NN | |
| 136.20 | |
| 172.66 | |
| 2-Ethylphenylhydrazine hydrochloride |
| ∼178°C (decomposition) | |
| C8H12N2 | |
| 50 g | |
| 3697547 | |
| JHPOWXCLWLEKBY-UHFFFAOYSA-N | |
| (2-ethylphenyl)hydrazine;hydrochloride | |
| 2733209 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H372-H351-H318-H317
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
H302-H317-H318-H351-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 421-460-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only