missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylphenylhydrazine hydrochloride, 97%

CAS: 19398-06-2 | C8H12N2 | 136.20 g/mol

Supplier:  Thermo Scientific Chemicals A1685330

 View more versions of this product

Catalog No. AAA1685330

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethylphenylhydrazine hydrochloride
∼178°C (decomposition)
C8H12N2
250 g
3697547
JHPOWXCLWLEKBY-UHFFFAOYSA-N
(2-ethylphenyl)hydrazine
2733209
98%
19398-06-2
1.21
MFCD00071599
UN3077
2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride
CCC1=CC=CC=C1NN
136.20
172.66

Chemical Identifiers

19398-06-2
136.20
JHPOWXCLWLEKBY-UHFFFAOYSA-N
2733209
C8H12N2
MFCD00071599
2-ethylphenylhydrazine hydrochloride, 2-ethylphenyl hydrazine hydrochloride, 2-ethylphenyl hydrazine hcl, 2-ethylphenylhydrazine, hcl, 1-2-ethylphenyl hydrazine hydrochloride, 2-ethyl phenyl hydrazine hydrochloride, 2-ethylphenylhydrazinehydrochloride, hydrazine, 2-ethylphenyl-, monohydrochloride, 2-ethylphenylhydrazine, chloride
CCC1=CC=CC=C1NN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H351-H318-H317
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye damage.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c

H302-H317-H318-H351-H372

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 421-460-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only