missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl salicylate, Thermo Scientific™

Catalog No. AC468830250
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468830250 25 mL
AC468831000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468830250 Supplier Thermo Scientific Chemicals Supplier No. 468830250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 118-60-5
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number 00053300
InChI Key FMRHJJZUHUTGKE-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Density 1.02
Flash Point 162°C
Physical Form Liquid
Color Colorless to Light Yellow
Melting Point -20°C
Boiling Point 190°C (21 mmHG)
Solubility Insoluble in water
Quantity 25 mL
Formula Weight 250.34
Chemical Name or Material 2-Ethylhexyl salicylate
Show More Show Less
ShelfLife 5 years
EINECSNumber 204-263-4
RTECSNumber VO3150000
Recommended Storage Normal conditions