missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Salicylate 98.0+%, TCI America™

Catalog No. S038725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S038725ML 25 mL
S0387500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S038725ML Supplier TCI America Supplier No. S038725ML

Chemical Identifiers

CAS 118-60-5
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.338
MDL Number MFCD00053300
InChI Key FMRHJJZUHUTGKE-UHFFFAOYSA-N
Synonym 2-ethylhexyl salicylate, octisalate, sunarome o, sunarome wmo, ethyl hexyl salicylate, benzoic acid, 2-hydroxy-, 2-ethylhexyl ester, usaf do-11, salicylic acid, 2-ethylhexyl ester, salicylic acid 2-ethylhexyl ester, ethylhexyl salicylate
PubChem CID 8364
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 250.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO3150000
TSCA Yes