missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol

Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol

$55.78 - $1064.48

Chemical Identifiers

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42303AK
View Documents
Thermo Scientific Chemicals
042303AK
250 mL
Each for $55.78
Only null left
 
AA42303K2
View Documents
Thermo Scientific Chemicals
042303K2
1 L
Each for $134.68
Only null left
 
AA42303K7
View Documents
Thermo Scientific Chemicals
042303K7
4 L
Each for $384.84
Only null left
 
AA42303M4
View Documents
Thermo Scientific Chemicals
042303M4
20 L
Each for $1,064.48
Only null left
 
Description

Description

Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

688-84-6
198.306
WDQMWEYDKDCEHT-UHFFFAOYSA-N
12713
CCCCC(CC)COC(=O)C(=C)C
C12H22O2
MFCD00009494
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
2-ethylhexyl 2-methylprop-2-enoate

Specifications

688-84-6
Colorless
218°C
Ether-like
1.438
250 mL
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
CCCCC(CC)COC(=O)C(=C)C
198.306
198.31
Liquid
-50°C
0.885 g/mL
92°C (198°F)
C12H22O2
MFCD00009494
1769420
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl 2-methylprop-2-enoate
12713
98%
2-Ethylhexyl methacrylate, Stabilized with 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H319-H335

EINECSNumber : 211-708-6

RTECSNumber : OZ4630000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only