Learn More
2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol
Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol
Supplier: Thermo Scientific Chemicals 042303K7
Description
Monomer used in the preparation of polymethacrylate polymers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Ethylhexyl methacrylate | |
| 688-84-6 | |
| Colorless | |
| 218°C | |
| Ether-like | |
| 1.438 | |
| 4 L | |
| 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391 | |
| CCCCC(CC)COC(=O)C(=C)C | |
| 198.306 | |
| 198.31 | |
| Liquid |
| Stabilized with 4-methoxyphenol | |
| -50°C | |
| 0.885 g/mL | |
| 92°C (198°F) | |
| C12H22O2 | |
| MFCD00009494 | |
| 1769420 | |
| WDQMWEYDKDCEHT-UHFFFAOYSA-N | |
| 2-ethylhexyl 2-methylprop-2-enoate | |
| 12713 | |
| 98% |
Chemical Identifiers
| 688-84-6 | |
| 198.306 | |
| WDQMWEYDKDCEHT-UHFFFAOYSA-N | |
| 12713 | |
| CCCCC(CC)COC(=O)C(=C)C |
| C12H22O2 | |
| MFCD00009494 | |
| 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391 | |
| 2-ethylhexyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319-H335
EINECSNumber : 211-708-6
RTECSNumber : OZ4630000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only