missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol

Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol

Supplier:  Thermo Scientific Chemicals 042303K2

 View more versions of this product

Catalog No. AA42303K2


Only null left
Explore available promotions

Description

Description

Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethylhexyl methacrylate
688-84-6
Colorless
218°C
Ether-like
1.438
1 L
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
CCCCC(CC)COC(=O)C(=C)C
198.306
198.31
Liquid
Stabilized with 4-methoxyphenol
-50°C
0.885 g/mL
92°C (198°F)
C12H22O2
MFCD00009494
1769420
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl 2-methylprop-2-enoate
12713
98%

Chemical Identifiers

688-84-6
198.306
WDQMWEYDKDCEHT-UHFFFAOYSA-N
12713
CCCCC(CC)COC(=O)C(=C)C
C12H22O2
MFCD00009494
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
2-ethylhexyl 2-methylprop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c

H227-H315-H317-H319-H335

EINECSNumber : 211-708-6

RTECSNumber : OZ4630000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only