missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexanoic Acid 99.0+%, TCI America™
$45.57 - $60.75
Chemical Identifiers
| CAS | 149-57-5 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.214 |
| MDL Number | MFCD00002675 |
| InChI Key | OBETXYAYXDNJHR-UHFFFAOYSA-N |
| Synonym | 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid |
| PubChem CID | 8697 |
| IUPAC Name | 2-ethylhexanoic acid |
| SMILES | CCCCC(CC)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
E012025ML
|
N/A | 25 mL |
Each for $45.57
|
|
|||||
|
E0120500ML
|
N/A | 500 mL |
Each for $60.75
|
|
|||||
Chemical Identifiers
| C8H16O2 | |
| MFCD00002675 | |
| 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid | |
| 2-ethylhexanoic acid |
Specifications
| 149-57-5 | |
| 228°C | |
| MFCD00002675 | |
| 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid | |
| CCCCC(CC)C(=O)O | |
| 144.214 | |
| 144.21 | |
| Liquid |
| Colorless | |
| C8H16O2 | |
| 3265 | |
| OBETXYAYXDNJHR-UHFFFAOYSA-N | |
| 2-ethylhexanoic acid | |
| 8697 | |
| ≥99.0% (GC) | |
| 2-Ethylhexanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : MO7700000
TSCA : Yes