Learn More
2-Ethylbutyric acid, 98%
CAS: 88-09-5 | C6H12O2 | 116.16 g/mol
$98.53 - $332.52
Chemical Identifiers
| CAS | 88-09-5 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00002670 |
| InChI Key | OXQGTIUCKGYOAA-UHFFFAOYSA-N |
| Synonym | 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid |
| PubChem CID | 6915 |
| IUPAC Name | 2-ethylbutanoic acid |
| SMILES | CCC(CC)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA16989AK
|
N/A | 250 mL |
Each for $98.53
|
|
|||||
|
AAA16989AU
|
N/A | 1000 mL |
Each for $332.52
|
|
|||||
Description
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.
Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
| C6H12O2 | |
| MFCD00002670 | |
| 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid | |
| 2-ethylbutanoic acid |
Specifications
| 88-09-5 | |
| 0.924 | |
| 87°C (188°F) | |
| 1.413 | |
| UN2922 | |
| 14,3110 | |
| Sparingly soluble with the usual organic solvents. Solubility in water 18g/L (20°C). | |
| CCC(CC)C(=O)O | |
| 116.16 | |
| 116.16 | |
| 2-Ethylbutyric acid |
| -15°C | |
| 192°C to 194°C | |
| C6H12O2 | |
| MFCD00002670 | |
| 1098634 | |
| 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid | |
| OXQGTIUCKGYOAA-UHFFFAOYSA-N | |
| 2-ethylbutanoic acid | |
| 6915 | |
| 98% |
Safety and Handling
GHS H Statement
H311-H314-H318-H227
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H312-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 201-796-4
RTECSNumber : ET1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only