missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyric acid, 98%

Catalog No. AAA16989AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16989AU 1000 mL
AAA16989AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16989AU Supplier Thermo Scientific Chemicals Supplier No. A16989AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-09-5 | C6H12O2 | 116.16 g/mol

Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.

Chemical Identifiers

CAS 88-09-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002670
InChI Key OXQGTIUCKGYOAA-UHFFFAOYSA-N
Synonym 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid
PubChem CID 6915
IUPAC Name 2-ethylbutanoic acid
SMILES CCC(CC)C(=O)O

Specifications

Melting Point -15°C
Density 0.924
Boiling Point 192°C to 194°C
Flash Point 87°C (188°F)
Refractive Index 1.413
Quantity 1000 mL
UN Number UN2922
Beilstein 1098634
Merck Index 14,3110
Solubility Information Sparingly soluble with the usual organic solvents. Solubility in water 18g/L (20°C).
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 2-Ethylbutyric acid
Show More Show Less
Hazard Category H227-H312-H314
Hazard Statement GHS H Statement
H311-H314-H318-H227
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 201-796-4
RTECSNumber ET1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only